siRNA Details
| Gene Symbol: Pfdn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013369.2 | NP_001013387.1 | 4 | 457 | ||
| NM_001110152.2 | NP_001103622.1 | 4 | 525 | |||
| NM_001199902.1 | NP_001186831.1 | 4 | 513 | |||
| GenBank mRNA | AK007940.1 | - | 4 | 318 | ||
| AK136249.1 | - | 4 | 526 | |||
| BC048510.1 | - | 4 | 297 | |||
| BC065407.1 | - | 4 | 457 | |||
| BC147673.1 | - | 4 | 358 | 9056646 | ||
| BC147674.1 | - | 4 | 358 | 9056647 | ||
| BU503885.1 | - | 3 | 296 | |||
| CB523981.1 | - | 2 | 133 | |||

Pfdn4
Genomic Map