siRNA Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010139.3 | NP_034269.2 | 7 | 1692 | ||
| GenBank mRNA | AK137704.1 | - | 7 | 1674 | ||
| AK144202.1 | - | 7 | 1676 | |||
| BC140960.1 | - | 7 | 1648 | 9055572 | ||
| U07634.1 | - | 7 | 1650 | |||
| X78339.1 | - | 7 | 1654 | |||

Epha2
Genomic Map