This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PLXNB2 SIRNA Assays ›
Gene Symbol:
Plxnb2
Gene Name:
plexin B2
Gene Aliases:
Debt, plexin-B2
Chromosome Location:
Chr.15: 89155546 - 89181140 on Build GRCm38
UniGene ID:
Mm.267514
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS274245
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plxnb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159521.2 NP_001152993.1 20 3318
NM_001284506.1 NP_001271435.1 19 3232
NM_138749.3 NP_620088.2 20 3282
GenBank mRNA AB093229.1 - 16 1867
AK147538.1 - 20 3283
AK154627.1 - 20 3233
BC007481.1 - 17 2080
BC036274.1 - 8 889
BC157960.1 - 20 3253 9088132
BC157961.1 - 20 3253 9088133
BC158086.1 - 20 3219 9088174
BC172162.1 - 20 3219 9054832

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
regulation of protein phosphorylation
signal transduction
multicellular organism development
neuroblast proliferation
brain development
regulation of cell shape
regulation of GTPase activity
positive regulation of axonogenesis
semaphorin-plexin signaling pathway
regulation of neuron migration
protein binding
semaphorin receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline