siRNA Details
| Gene Symbol: Gabpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008065.2 | NP_032091.2 | 5 | 900 | ||
| GenBank mRNA | AK139916.1 | - | 5 | 580 | ||
| AK145446.1 | - | 5 | 580 | |||
| AK145838.1 | - | 5 | 645 | |||
| AK148087.1 | - | 5 | 671 | |||
| AK150455.1 | - | 5 | 580 | |||
| BC013562.1 | - | 3 | 295 | 3493987 | ||
| BC052448.1 | - | 5 | 900 | 6408997 | ||
| M74515.1 | - | 5 | 907 | |||

Gabpa
Genomic Map