siRNA Details
| Gene Symbol: Hoxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010455.2 | NP_034585.1 | 2 | 1721 | ||
| GenBank mRNA | AK033500.1 | - | 2 | 623 | ||
| BC036986.1 | - | 4 | 688 | |||
| BC131978.1 | - | 2 | 585 | 40130255 | ||
| BC132643.1 | - | 2 | 585 | 40130920 | ||
| M17192.1 | - | 2 | 653 | |||

Hoxa7
Genomic Map