siRNA Details
| Gene Symbol: Mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081979.2 | NP_001075448.1 | 2 | 514 | ||
| NM_010788.4 | NP_034918.1 | 3 | 638 | |||
| GenBank mRNA | AB221662.1 | - | 2 | 359 | ||
| AF072251.1 | - | 2 | 359 | |||
| AF158181.1 | - | 3 | 541 | |||
| AJ132922.1 | - | 2 | 394 | |||
| AK158664.1 | - | 2 | 461 | |||
| BC027153.1 | - | 3 | 560 | 4948925 | ||

Mecp2
Genomic Map