This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSME1 SIRNA Assays ›
Gene Symbol:
Psme1
Gene Name:
proteasome (prosome, macropain) activator subunit 1 (PA28 alpha)
Gene Aliases:
AW413925, PA28a
Chromosome Location:
Chr.14: 55578494 - 55581527 on Build GRCm38
UniGene ID:
Mm.830
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS276519
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psme1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011189.1 NP_035319.1 3 165
GenBank mRNA AK008514.1 - 3 140
AK019390.1 - 3 193
AK150708.1 - 3 209
AK150975.1 - 3 208
BC035549.1 - 2 323
BC088981.1 - 3 185 5689048
D87909.1 - 3 165
U60328.1 - 3 84

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017264, FQ226855, FQ230622, BC060574, FQ224986, and others.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endopeptidase activity
antigen processing and presentation of exogenous antigen
protein binding
endopeptidase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline