siRNA Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122829.1 | NP_001116301.1 | 4 | 807 | ||
| NM_030680.2 | NP_109605.2 | 4 | 807 | |||
| GenBank mRNA | AF182947.1 | - | 5 | 796 | ||
| AF322655.1 | - | 4 | 527 | |||
| AK148196.1 | - | 5 | 784 | |||
| BC052149.1 | - | 4 | 745 | 6332288 | ||
| BC056442.1 | - | 4 | 807 | 5702493 | ||

Upf1
Genomic Map