This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SKP1A SIRNA Assays ›
Gene Symbol:
Skp1a
Gene Name:
S-phase kinase-associated protein 1A
Gene Aliases:
15kDa, 2610043E24Rik, 2610206H23Rik, EMC19, OCP-II, OCP2, SKP1, Tceb1l, p19A, p19Skp1
Chromosome Location:
Chr.11: 52230522 - 52246858 on Build GRCm38
UniGene ID:
Mm.42944
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS277839
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Skp1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011543.4 NP_035673.3 4, 5 438
GenBank mRNA AF083214.1 - 4, 5 362
AK002983.1 - 4, 5 409
AK010628.1 - 4, 5 424
AK012498.1 - 4, 5 397
AK014245.1 - 4, 5 397
AK027909.1 - 4, 5 396
AK078330.1 - 4, 5 424
AK088339.1 - 4, 5 397
AK152609.1 - 4, 5 396
AK160198.1 - 4, 5 397
AK166673.1 - 4, 5 397
BC002115.1 - 4, 5 381 3490159
BU841654.1 - 5, 6 530
Z47088.1 - 4, 5 387

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001007608, XM_006246244, FQ212502, FQ212745, FQ214369, and others.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
histone H2A monoubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0