siRNA Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175443.5 | NP_780652.2 | 6 | 1181 | ||
| GenBank mRNA | AK077179.1 | - | 6 | 920 | ||
| AK087818.1 | - | 5 | 979 | |||
| AY212244.1 | - | 6 | 772 | |||
| BC152310.1 | - | 6 | 1119 | 40089976 | ||

Etnk2
Genomic Map