siRNA Details
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175450.4 | NP_780659.2 | 4 | 484 | ||
GenBank mRNA | AK050106.1 | - | 4 | 482 | ||
AK082695.1 | - | 4 | 486 | |||
AK133716.1 | - | 4 | 485 | |||
AK160713.1 | - | 4 | 482 | |||
AK166113.1 | - | 482 | ||||
AK168691.1 | - | 4 | 482 | |||
BC032968.1 | - | 3 | 176 | |||
BC095984.1 | - | 4 | 484 | 4978914 |
