siRNA Details
| Gene Symbol: Trib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144551.5 | NP_653134.2 | 3 | 2057 | ||
| GenBank mRNA | AK044747.1 | - | 3 | 2015 | ||
| AK080064.1 | - | 3 | 1260 | |||
| AK082329.1 | - | 3 | 1269 | |||
| AK194281.1 | - | 1 | 65 | |||
| BC037387.1 | - | 3 | 2001 | 5363174 | ||

Trib2
Genomic Map