This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CEP170B SIRNA Assays ›
Gene Symbol:
Cep170b
Gene Name:
centrosomal protein 170B
Gene Aliases:
AI466840, AW555464, Kiaa0284, mKIAA0284
Chromosome Location:
Chr.12: 112720591 - 112746592 on Build GRCm38
UniGene ID:
Mm.23689
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS278298
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep170b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024602.3 NP_001019773.2 12 2417
GenBank mRNA AK122236.1 - 13 2346
AK147410.1 - 12 2358
AK147612.1 - 10 2022
BC141174.1 - 11 2217 9055786

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline