siRNA Details
| Gene Symbol: BC051665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199148.2 | NP_954599.2 | 5 | 685 | ||
| GenBank mRNA | AK145559.1 | - | 5 | 683 | ||
| BC051665.1 | - | 5 | 670 | 30027545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC051665| Gene Symbol: BC051665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199148.2 | NP_954599.2 | 5 | 685 | ||
| GenBank mRNA | AK145559.1 | - | 5 | 683 | ||
| BC051665.1 | - | 5 | 670 | 30027545 | ||