This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TSKS SIRNA Assays ›
Gene Symbol:
Tsks
Gene Name:
testis-specific serine kinase substrate
Gene Aliases:
Stk22s1, Tssks1
Chromosome Location:
Chr.7: 44929577 - 44958035 on Build GRCm38
UniGene ID:
Mm.84502
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS278564
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tsks
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077591.1 NP_001071059.1 5 630
NM_011651.2 NP_035781.2 5 657
GenBank mRNA AF025310.2 - 5 637
BC118618.1 - 5 609 40107271
BC121827.1 - 5 609 40107270
CN831975.1 - 380

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199088, XM_006229014, XM_006229016, XM_006229017, XM_006229019, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4m7g7:80/100.66.75.155:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline