This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM191C SIRNA Assays ›
Gene Symbol:
Tmem191c
Gene Name:
transmembrane protein 191C
Gene Aliases:
4933405M22Rik, D16Bwg1494e, MNCb-4137
Chromosome Location:
Chr.16: 17276300 - 17278661 on Build GRCm38
UniGene ID:
Mm.246388
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS278799
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem191c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177473.3 NP_803424.2 6 524
GenBank mRNA AB041601.1 - 4 715
BC058760.1 - 2 619
BC098238.1 - 2 151
BU936413.1 - 6 507
BY296351.1 - 6 379

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006248711, XM_001059247.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline