This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other D17WSU92E SIRNA Assays ›
Gene Symbol:
D17Wsu92e
Gene Name:
DNA segment, Chr 17, Wayne State University 92, expressed
Gene Aliases:
AU020189, C030010A15, C80239
Chromosome Location:
Chr.17: 27751232 - 27820542 on Build GRCm38
UniGene ID:
Mm.196577
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS278894
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: D17Wsu92e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033279.3 NP_001028451.2 4 775
NM_001044719.2 NP_001038184.1 4 775
GenBank mRNA AK143016.1 - 4 687
AK161603.1 - 4 776
AK165011.1 - 4 775
BC132324.1 - 5 701 40130601
BC132326.1 - 5 701 40130603
BC137573.1 - 4 775 8860592
BC137576.1 - 4 775 8860595
BC145403.1 - 4 775 9053961
BU922193.1 - 3 373
CF536829.1 - 1 28

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_022758, NM_024294, BC010184, BC075810, AK056964, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
macroautophagy
molecular_function
ubiquitin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0