siRNA Details
| Gene Symbol: Gm4861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177665.3 | NP_808333.1 | 3 | 381 | ||
| GenBank mRNA | AK079008.1 | - | 3 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4861| Gene Symbol: Gm4861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177665.3 | NP_808333.1 | 3 | 381 | ||
| GenBank mRNA | AK079008.1 | - | 3 | 382 | ||