siRNA Details
| Gene Symbol: Ak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081277.1 | NP_001074746.1 | 13 | 1729 | ||
| GenBank mRNA | AB060081.1 | - | 4 | 341 | ||
| AK053807.1 | - | 13 | 1727 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ak5| Gene Symbol: Ak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081277.1 | NP_001074746.1 | 13 | 1729 | ||
| GenBank mRNA | AB060081.1 | - | 4 | 341 | ||
| AK053807.1 | - | 13 | 1727 | |||