siRNA Details
| Gene Symbol: Cyp2j13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145548.4 | NP_663523.2 | 7 | 1197 | ||
| GenBank mRNA | AK143903.1 | - | 7 | 1198 | ||
| AK143981.1 | - | 7 | 1194 | |||
| AK162358.1 | - | 1201 | ||||
| AK165405.1 | - | 7 | 1197 | |||
| AK165469.1 | - | 7 | 1175 | |||
| AK165473.1 | - | 7 | 1175 | |||
| BC016446.1 | - | 6 | 1010 | 4235131 | ||
| BC024451.1 | - | 6 | 817 | |||

Cyp2j13
Genomic Map