siRNA Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172277.2 | NP_758481.1 | 8, 9 | 984 | ||
| GenBank mRNA | AK045061.1 | - | 8, 9 | 977 | ||
| AK082871.1 | - | 8, 9 | 977 | |||
| AK148429.1 | - | 8, 9 | 977 | |||
| AK156817.1 | - | 8, 9 | 930 | |||
| AK161672.1 | - | 7, 8 | 918 | |||
| BC019142.1 | - | 7, 8 | 980 | |||
| BC037599.1 | - | 8, 9 | 1010 | 5150754 | ||

Snx8
Genomic Map