siRNA Details
| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081267.2 | NP_001074736.1 | 6 | 1593 | ||
| GenBank mRNA | AK032853.1 | - | 6 | 1569 | ||
| AK146675.1 | - | 6 | 1603 | |||
| BC085127.1 | - | 6 | 1594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsf1| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081267.2 | NP_001074736.1 | 6 | 1593 | ||
| GenBank mRNA | AK032853.1 | - | 6 | 1569 | ||
| AK146675.1 | - | 6 | 1603 | |||
| BC085127.1 | - | 6 | 1594 | |||