This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM362 SIRNA Assays ›
Gene Symbol:
Gm362
Gene Name:
predicted gene 362
Gene Aliases:
-
Chromosome Location:
Chr.X: 43590989 - 43593530 on Build GRCm38
UniGene ID:
Mm.368463
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS280101
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm362
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195271.1 NP_001182200.1 1 1521

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-cdc4cf686-q6tt6:80/100.66.77.81:80.
git-commit: 18f7f941bc72f70e8ff4e0be257f7363f2314623
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.0-Offline