siRNA Details
| Gene Symbol: Fbll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004147.3 | NP_001004147.1 | 1 | 1328 | ||
| GenBank mRNA | AK140117.1 | - | 1 | 1034 | ||
| BC052068.1 | - | 1 | 1018 | 5710336 | ||
| BU701095.1 | - | 1 | 191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbll1| Gene Symbol: Fbll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004147.3 | NP_001004147.1 | 1 | 1328 | ||
| GenBank mRNA | AK140117.1 | - | 1 | 1034 | ||
| BC052068.1 | - | 1 | 1018 | 5710336 | ||
| BU701095.1 | - | 1 | 191 | |||