siRNA Details
| Gene Symbol: Gpr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033360.3 | NP_001028532.1 | 2 | 1451 | ||
| GenBank mRNA | AK138185.1 | - | 2 | 1395 | ||
| AK138370.1 | - | 2 | 1453 | |||
| BC139013.1 | - | 1 | 1243 | 8862035 | ||
| BC139014.1 | - | 1 | 1243 | 8862036 | ||
| DQ198079.1 | - | 2 | 1411 | |||
More Information
Set Membership: |
|

Gpr101
Genomic Map