This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR644 SIRNA Assays ›
Gene Symbol:
Olfr644
Gene Name:
olfactory receptor 644
Gene Aliases:
MOR13-1
Chromosome Location:
Chr.7: 104068085 - 104069029 on Build GRCm38
UniGene ID:
Mm.377862
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS281810
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr644
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147121.1 NP_667332.1 1 546
GenBank mRNA BC099886.1 - 1 538
BC099887.1 - 1 538

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline