This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VAT1 SIRNA Assays ›
Gene Symbol:
Vat1
Gene Name:
vesicle amine transport protein 1 homolog (T californica)
Gene Aliases:
VAT-1
Chromosome Location:
Chr.11: 101458748 - 101466199 on Build GRCm38
UniGene ID:
Mm.29210
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS282119
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vat1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012037.2 NP_036167.1 2 678
GenBank mRNA AK034795.1 - 2 680
AK154965.1 - 2 661
AK159310.1 - 2 621
AK159315.1 - 2 663
AK160891.1 - 2 670
AK170371.1 - 2 659
AK181346.1 - 2 233
AK188590.1 - 11
AK199541.1 - 1 159
AK207392.1 - 11
BC085504.1 - 2 597
BC099678.1 - 2 660 6311681

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mitochondrial fusion
oxidation-reduction process
zinc ion binding
N-ethylmaleimide reductase activity
oxidoreductase activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
epoxyqueuosine reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0