siRNA Details
| Gene Symbol: Tmtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177368.4 | NP_796342.2 | 5, 6 | 2412 | ||
| GenBank mRNA | AK052344.1 | - | 5, 6 | 2385 | ||
| BC090655.1 | - | 5, 6 | 1774 | |||
| BC092226.1 | - | 5, 6 | 2368 | 6852756 | ||
| BC138362.1 | - | 5, 6 | 2143 | 8861382 | ||
| BC138363.1 | - | 5, 6 | 2143 | 8861383 | ||

Tmtc2
Genomic Map