This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPATA33 SIRNA Assays ›
Gene Symbol:
Spata33
Gene Name:
spermatogenesis associated 33
Gene Aliases:
4732415M23Rik, AI847264
Chromosome Location:
Chr.8: 123212816 - 123222045 on Build GRCm38
UniGene ID:
Mm.258354
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS283014
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spata33
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177279.4 NP_796253.2 2 165
GenBank mRNA AK028607.1 - 1 282
AK049895.1 - 1 245
AK053679.1 - 1 45
AK076656.1 - 2 141
BC059919.1 - 1 230 6827099

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0