siRNA Details
| Gene Symbol: Sfmbt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198808.1 | NP_001185737.1 | 6 | 676 | ||
| NM_001198809.1 | NP_001185738.1 | 6 | 755 | |||
| NM_177386.5 | NP_796360.2 | 7 | 799 | |||
| GenBank mRNA | AK040600.1 | - | 6 | 756 | ||
| AK049505.1 | - | 6 | 618 | |||
| AK052339.1 | - | 6 | 742 | |||
| AK147557.1 | - | 6 | 681 | |||
| AK147651.1 | - | 7 | 654 | |||
| AK168444.1 | - | 8 | 704 | |||
| AK220408.1 | - | 4 | 541 | |||
| BC115510.1 | - | 6 | 610 | |||
| BC115511.1 | - | 6 | 610 | |||
| BC139354.1 | - | 6 | 587 | 8862376 | ||
| BC145610.1 | - | 5 | 543 | 9054176 | ||

Sfmbt2
Genomic Map