siRNA Details
| Gene Symbol: Dkkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015789.3 | NP_056604.2 | 5 | 823 | ||
| GenBank mRNA | AF177399.1 | - | 5 | 728 | ||
| AF274312.1 | - | 5 | 782 | |||
| AK131694.1 | - | 5 | 768 | |||
| BC002215.1 | - | 5 | 801 | 3490482 | ||
| BE134426.1 | - | 1 | 81 | |||

Dkkl1
Genomic Map