siRNA Details
| Gene Symbol: Gm6034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034909.3 | NP_001030081.2 | 3 | 393 | ||
| GenBank mRNA | AK156795.1 | - | 394 | |||
| BC147141.1 | - | 3 | 373 | 9056198 | ||
| BC147142.1 | - | 3 | 373 | 9056188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6034| Gene Symbol: Gm6034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034909.3 | NP_001030081.2 | 3 | 393 | ||
| GenBank mRNA | AK156795.1 | - | 394 | |||
| BC147141.1 | - | 3 | 373 | 9056198 | ||
| BC147142.1 | - | 3 | 373 | 9056188 | ||