siRNA Details
| Gene Symbol: Hebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019487.3 | NP_062360.1 | 2 | 460 | ||
| GenBank mRNA | AF117614.1 | - | 2 | 390 | ||
| AK038452.1 | - | 2 | 449 | |||
| AK078130.1 | - | 2 | 460 | |||
| BC006898.1 | - | 2 | 271 | 3599858 | ||

Hebp2
Genomic Map