siRNA Details
| Gene Symbol: St7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083315.2 | NP_001076784.1 | 3, 4 | 686 | ||
| NM_001289624.1 | NP_001276553.1 | 3, 4 | 610 | |||
| NM_001289625.1 | NP_001276554.1 | 3, 4 | 610 | |||
| NM_001289626.1 | NP_001276555.1 | 3, 4 | 610 | |||
| NM_001289627.1 | NP_001276556.1 | 3, 4 | 344 | |||
| NM_001289629.1 | NP_001276558.1 | 3, 4 | 344 | |||
| NM_022332.3 | NP_071727.2 | 3, 4 | 686 | |||
| GenBank mRNA | AF234884.1 | - | 3, 4 | 637 | ||
| AF234885.1 | - | 3, 4 | 637 | |||
| AK079107.1 | - | 3, 4 | 687 | |||
| AK132103.1 | - | 3, 4 | 640 | |||
| AK151550.1 | - | 3, 4 | 641 | |||
| AY007792.1 | - | 3, 4 | 572 | |||
| AY007793.1 | - | 3, 4 | 572 | |||
| AY007794.1 | - | 4, 5 | 601 | |||
| AY007795.1 | - | 4, 5 | 601 | |||
| AY007796.1 | - | 4, 5 | 411 | |||
| AY007797.1 | - | 4, 5 | 411 | |||
| AY007798.1 | - | 3, 4 | 344 | |||
| AY007799.1 | - | 3, 4 | 344 | |||
| AY551821.1 | - | 3, 4 | 337 | |||
| AY551822.1 | - | 3, 4 | 337 | |||
| BC024652.1 | - | 3, 4 | 610 | 3660309 | ||
| BC051429.1 | - | 3, 4 | 464 | |||
| BC060630.1 | - | 3, 4 | 527 | |||
| BC108418.1 | - | 3, 4 | 630 | 30603484 | ||
| CT010245.1 | - | 3, 4 | 263 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001004102, XM_006236135, XM_008762701, XM_006236132, XM_008762700, and others.

St7
Genomic Map