siRNA Details
| Gene Symbol: Gcnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039560.3 | NP_001034649.1 | 2 | 205 | ||
| GenBank mRNA | AK039414.1 | - | 2 | 207 | ||
| BC139073.1 | - | 2 | 165 | 8862095 | ||
| BC139076.1 | - | 2 | 165 | 8862098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcnt7| Gene Symbol: Gcnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039560.3 | NP_001034649.1 | 2 | 205 | ||
| GenBank mRNA | AK039414.1 | - | 2 | 207 | ||
| BC139073.1 | - | 2 | 165 | 8862095 | ||
| BC139076.1 | - | 2 | 165 | 8862098 | ||