siRNA Details
| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025317.2 | NP_079593.1 | 1 | 132 | ||
| GenBank mRNA | AK002348.1 | - | 1 | 134 | ||
| AK003225.1 | - | 1 | 133 | |||
| AK009657.1 | - | 1 | 115 | |||
| BC056985.1 | - | 1 | 115 | 6818054 | ||

Mrpl54
Genomic Map