This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CNPY4 SIRNA Assays ›
Gene Symbol:
Cnpy4
Gene Name:
canopy 4 homolog (zebrafish)
Gene Aliases:
2610019P18Rik
Chromosome Location:
Chr.5: 138187535 - 138193894 on Build GRCm38
UniGene ID:
Mm.24592
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS288103
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnpy4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178612.4 NP_848727.1 4 440
GenBank mRNA AK011474.1 - 4 441
AK012968.1 - 4 422
AK033219.1 - 4 442
AK051562.1 - 4 442
BC086647.1 - 4 405 6818372
BC094672.1 - 4 547 30633103

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein localization
receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline