This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC16A9 SIRNA Assays ›
Gene Symbol:
Slc16a9
Gene Name:
solute carrier family 16 (monocarboxylic acid transporters), member 9
Gene Aliases:
1200003C15Rik, 4930425B13Rik, AI552636
Chromosome Location:
Chr.10: 70245276 - 70285951 on Build GRCm38
UniGene ID:
Mm.19325
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS288801
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc16a9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025807.3 NP_080083.2 5 621
GenBank mRNA AK004556.1 - 5 622
AK004599.1 - 5 800
AK004684.2 - 5 679
AK015196.1 - 3 398
BC034205.1 - 5 576 4987055
BC055839.1 - 5 579 4237811

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
monocarboxylic acid transport
ion transmembrane transport
urate metabolic process
transmembrane transport
anion transmembrane transport
organic acid transmembrane transport
molecular_function
monocarboxylic acid transmembrane transporter activity
symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-rs9q2:80/100.66.78.195:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline