siRNA Details
| Gene Symbol: Pqlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025861.3 | NP_080137.2 | 4, 5 | 617 | ||
| GenBank mRNA | AK018759.1 | - | 3, 4 | 995 | ||
| BC043686.1 | - | 795 | 5135928 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pqlc1| Gene Symbol: Pqlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025861.3 | NP_080137.2 | 4, 5 | 617 | ||
| GenBank mRNA | AK018759.1 | - | 3, 4 | 995 | ||
| BC043686.1 | - | 795 | 5135928 | |||