siRNA Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025909.3 | NP_080185.1 | 3 | 640 | ||
| GenBank mRNA | AK008020.1 | - | 3 | 641 | ||
| AK076209.1 | - | 3 | 670 | |||
| AK136208.1 | - | 3 | 666 | |||
| BC016238.1 | - | 2 | 595 | 4504835 | ||

Oma1
Genomic Map