siRNA Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026032.1 | NP_080308.1 | 2, 3 | 193 | ||
| GenBank mRNA | AK041290.1 | - | 2, 3 | 193 | ||
| AK048497.1 | - | 2, 3 | 226 | |||
| BC021460.1 | - | 2, 3 | 476 | 5054997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm1| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026032.1 | NP_080308.1 | 2, 3 | 193 | ||
| GenBank mRNA | AK041290.1 | - | 2, 3 | 193 | ||
| AK048497.1 | - | 2, 3 | 226 | |||
| BC021460.1 | - | 2, 3 | 476 | 5054997 | ||