siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF281634.1 | - | 2 | 452 | ||
| AK082970.1 | - | 2 | 355 | |||
| AK152681.1 | - | 2 | 339 | |||
| AK169251.1 | - | 2 | 601 | |||
| AK178961.1 | - | 1 | 64 | |||
| AK212218.1 | - | 1 | 64 | |||
| AK214724.1 | - | 1 | 64 | |||
| BC018339.1 | - | 1 | 324 | 3583940 | ||

Zfp422
Genomic Map