siRNA Details
| Gene Symbol: Ufm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026435.5 | NP_080711.1 | 2, 3 | 109 | ||
| GenBank mRNA | AK007788.1 | - | 2, 3 | 73 | ||
| AK008280.1 | - | 2, 3 | 72 | |||
| AK011737.1 | - | 2, 3 | 90 | |||
| AK089948.1 | - | 2, 3 | 75 | |||
| AK148146.1 | - | 2, 3 | 79 | |||
| AK166332.1 | - | 2, 3 | 103 | |||
| AK167301.1 | - | 2, 3 | 78 | |||
| AK167321.1 | - | 2, 3 | 73 | |||
| BC051554.1 | - | 1, 2 | 73 | |||
| BC061065.1 | - | 2, 3 | 108 | 6594260 | ||
| CJ075089.1 | - | 2, 3 | 135 | |||

Ufm1
Genomic Map