siRNA Details
| Gene Symbol: Dusp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024438.4 | NP_077758.1 | 1 | 251 | ||
| GenBank mRNA | AB038769.1 | - | 1 | 206 | ||
| AB051896.1 | - | 1 | 225 | |||
| AF237618.1 | - | 1 | 423 | |||
| AK010127.1 | - | 1 | 248 | |||
| AK049927.1 | - | 1 | 216 | |||
| AK077871.1 | - | 1 | 252 | |||
| AK083610.1 | - | 200 | ||||
| BC021591.1 | - | 1 | 206 | 5149051 | ||

Dusp19
Genomic Map