This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NCAPD2 SIRNA Assays ›
Gene Symbol:
Ncapd2
Gene Name:
non-SMC condensin I complex, subunit D2
Gene Aliases:
2810406C15Rik, 2810465G24Rik, CAP-D2, CNAP1, mKIAA0159
Chromosome Location:
Chr.6: 125168007 - 125191668 on Build GRCm38
UniGene ID:
Mm.257590
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290170
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ncapd2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146171.1 NP_666283.1 3 293
GenBank mRNA AK129068.1 - 3 258
AK156143.1 - 3 323
BC025460.1 - 3 313
BC026982.1 - 3 293 5364933
BC029133.1 - 3 250 4951596

Back To Top

More Information


Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
mitotic chromosome condensation
chromosome condensation
cell division
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0