This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700008O03RIK SIRNA Assays ›
Gene Symbol:
1700008O03Rik
Gene Name:
RIKEN cDNA 1700008O03 gene
Gene Aliases:
-
Chromosome Location:
Chr.7: 44358843 - 44385692 on Build GRCm38
UniGene ID:
Mm.358922
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290670
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700008O03Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027049.1 NP_081325.1 2, 3 248
GenBank mRNA AK005784.1 - 2, 3 249
BC127243.1 - 1, 2 194
BC127244.1 - 1, 2 194

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline