This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2310009B15RIK SIRNA Assays ›
Gene Symbol:
2310009B15Rik
Gene Name:
RIKEN cDNA 2310009B15 gene
Gene Aliases:
-
Chromosome Location:
Chr.1: 138851979 - 138856854 on Build GRCm38
UniGene ID:
Mm.264673
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290807
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2310009B15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081226.1 NP_001074695.1 2, 3 382
GenBank mRNA AK009246.1 - 2, 3 555

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108347, BC166874, BC166924, BC166876.

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline