siRNA Details
| Gene Symbol: 1700028J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006462.1 | - | 2, 3 | 254 | ||
| BC049653.1 | - | 2, 3 | 219 | |||
| BC061239.1 | - | 1, 2 | 134 | |||
| BC087938.1 | - | 1, 2 | 134 | |||
| BC100477.1 | - | 2, 3 | 235 | |||

1700028J19Rik
Genomic Map