This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DOPEY2 SIRNA Assays ›
Gene Symbol:
Dopey2
Gene Name:
dopey family member 2
Gene Aliases:
0610038M01Rik, 2610510B01Rik, AI503246, C21orf5, mKIAA0933
Chromosome Location:
Chr.16: 93711907 - 93810588 on Build GRCm38
UniGene ID:
Mm.23230
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS291053
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dopey2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026700.2 NP_080976.1 3 418
NM_027293.1 NP_081569.1 3 418
GenBank mRNA AK002808.1 - 3 348
AK147255.1 - 3 396
AK147536.1 - 2 438
AK156170.1 - 3 392
AK164839.1 - 3 395

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
cognition
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0