This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ATP6AP2 SIRNA Assays ›
Gene Symbol:
Atp6ap2
Gene Name:
ATPase, H+ transporting, lysosomal accessory protein 2
Gene Aliases:
(P)RR, 5730403E06Rik, APT6M8-9, ATP6M8-9, Atp6ip2, M8-9
Chromosome Location:
Chr.X: 12587759 - 12617051 on Build GRCm38
UniGene ID:
Mm.25148
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS291221
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price Contact Us
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp6ap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027439.4 NP_081715.1 8 926
GenBank mRNA AB192471.2 - 8 802
AK017482.1 - 8 858
AK029405.1 - 8 829
AK076495.1 - 8 871
AK151735.1 - 8 858
AK168139.1 - 8 883
AW985491.1 - 5 493
BC014706.1 - 8 797 4457079

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiotensin maturation
rostrocaudal neural tube patterning
positive regulation of Wnt signaling pathway
positive regulation of transforming growth factor beta1 production
regulation of MAPK cascade
eye pigmentation
head morphogenesis
aspartic-type endopeptidase activity
receptor activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-nrgn4:80/100.66.79.50:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline